Orders LinkContactUsLinkComponent
Azotobacter chroococcum Slide
Azotobacter chroococcum, Smear, Gram (-)
  470180-860
 :   Avantor
Azotobacter chroococcum Slide
  470180-860
 :  Avantor
Azotobacter chroococcum Slide
Azotobacter chroococcum Slide
Azotobacter chroococcum, Smear, Gram (-)
$135.18
Each (1UN)
Microaerophilic Bacterium
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Rods in pairs, packets, and chains. Nitrogen fixer in soil.
  • Description:
    Azotobacter chroococcum
  • Stain:
    Gram (-)
  • Orientation:
    Smear