Azotobacter chroococcum Slide
Azotobacter chroococcum, Smear, Gram (-)
470180-860
: Avantor
Microaerophilic Bacterium
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rods in pairs, packets, and chains. Nitrogen fixer in soil.
- Description:Azotobacter chroococcum
- Stain:Gram (-)
- Orientation:Smear