Auerbach’s Plexus, section, azan stained slides
Auerbach’s Plexus, Section, Azan
Catalog # 470182-704
Supplier: Avantor
Jump to:
Product Details & Documents
Innervation within the Gut
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal intestine. Clusters of parasympathetic ganglion cells.
Especificaciones
- Description:Auerbach’s Plexus
- Stain:Azan
- Orientation:Section