Aspirator Filter Pump
Aspirator Filter Pump
Catalog # 470202-842
Supplier: HUMBOLDT MFG CO
Número de proveedor #: H-12020
Jump to:
Product Details & Documents
Ideal for vacuum filtrations under both high and low water pressure.
- A ball-type, positive action valve in the suction tube prevents backflow if pressure is reduced
- Nickel-plated and designed for greater air entrainment capacity
- Chapman, Swirling-type
- 3/8" NPT tapered
- Size: 5.375" L
Especificaciones
- Description:Aspirator Filter Pump
- Cat. No.:470202-842