Orders LinkContactUsLinkComponent
Ward's® Artificial Urine
Ward's® Artificial Urine
Supplier:   ALDON CORP SE
Ward's® Artificial Urine
Supplier:  ALDON CORP SE
Certificates
Ward's® Artificial Urine
Ward's® Artificial Urine
4 Options Available Below
Product Details & Documents
Completely safe and realistic for urinalysis.
Students may find urology testing demonstrations or lab exercises more pleasant if they do not have to use the real thing.
Caution: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Product Family Options
Información del producto
  • Grade
    Size
    Availability
    Price
  • 470024-546
    Artificial urine control
    Artificial urine control
    Laboratory
    500 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $272.37
    $272.37
    Each (1UN)
       $272.37
    Supplier Number: UU0075-500ML
    Especificaciones:
    More
    Specifications
    Cat. No.
    470024-546
  • 470024-548
    Artificial urine with phosphates
    Artificial urine with phosphates
    Laboratory
    500 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $287.28
    $287.28
    Each (1UN)
       $287.28
    Supplier Number: UU0085
    Especificaciones:
    More
    Specifications
    Cat. No.
    470024-548
  • 470024-550
    Artificial urine with glucose
    Artificial urine with glucose
    Laboratory
    500 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $259.03
    $259.03
    Each (1UN)
       $259.03
    Supplier Number: UU0080-500
    Especificaciones:
    More
    Specifications
    Cat. No.
    470024-550
  • 470024-552
    Artificial urine with albumin
    Artificial urine with albumin
    Laboratory
    500 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $376.83
    $376.83
    Each (1UN)
       $376.83
    Supplier Number: UU0070-500
    Especificaciones:
    More
    Specifications
    Cat. No.
    470024-552