Areolar Tissue Slide
Areolar Tissue, Whole mount, Verhoeff and Eosin
Catalog # 470177-570
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates specific tissue characteristics
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Spread film of fasciae. Collagenous and elastic fibers.
Especificaciones
- Description:Areolar Tissue
- Stain:Verhoeff and Eosin
- Orientation:Whole mount