Arcella, Whole Mount Slides
Arcella, Explano-Mount™
Catalog # 470181-452
Supplier: Avantor
Jump to:
Product Details & Documents
Hard shelled Amoeba
- Identifying features clearly distinguishable
- Rigorous quality control standards
An amoeboid protist with a chitinous ornamental test (hard shell) and central aperture.
Especificaciones
- Description:Arcella
- Orientation:Explano-Mount™