Orders LinkContactUsLinkComponent
Big Books
Big Books
Seeds Get Around Student Book (Nancy White), 15 cm, 20 cm
Catalog # 470047-860
Supplier:   SUNDANCE/NEWBRIDGE
Supplier #:  825262
Big Books
Catalog # 470047-860
Supplier:  SUNDANCE/NEWBRIDGE
Big Books
Big Books
Seeds Get Around Student Book (Nancy White), 15 cm, 20 cm
Price
$839.50
Log in for availability
Each (1UN)
Product Details & Documents
Brilliantly bring nature to life with the vivid images and simple text of these award-winning early science Big Books. Each oversized book measures 14×20", and is large enough for students to see from the back of the classroom.
Student versions allow your class to follow along at their desks while you read. All Big Books meet National Science Standards and benchmarks for science literacy. Choose from a wide variety of titles - from animals and weather to electricity and ecology. Nothing is cuter than a baby animal. Show your students images of various animal infants being cared for and illustrate how animal mothers provide food, warmth, protection, transportation, and cleansing for their young.
Specifications
  • Description:
    Seeds Get Around Student Book (Nancy White)
  • Length:
    15 cm
  • Width:
    20 cm