ContactUsLinkComponent
Analysis Of Restriction Enzyme Clevage Patterns Of DNA Lab Activity Kit
Analysis Of Restriction Enzyme Clevage Patterns Of DNA Lab Activity Kit
Catalog # 470007-374
Supplier:  EDVOTEK, INC.
undefined
Analysis Of Restriction Enzyme Clevage Patterns Of DNA Lab Activity Kit
Catalog # 470007-374
Supplier:  EDVOTEK, INC.
Supplier Number:  102
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Descripción:
    Analysis Of Restriction Enzyme Clevage Patterns Of DNA Lab Activity Kit
  • Referencia:
    470007-374
  • Nº de proveedor:
    102
Specifications
About this item
Restriction enzymes are endonucleases that recognize and cut double-stranded DNA within or near defined base sequences. They are essential tools for biotechnology.
Experiment contains several DNAs digested with various restriction enzymes to demonstrate patterns and frequency of cleavage. Digests are separated by agarose gel electrophoresis.
Include materials for six student groups to run six gels of six lanes each. Requires, but does not include horizontal gel electrophoresis apparatus, DC power supply, and automatic micropipets with tips.