Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Descripción:Analysis of Minerals and Soils Lab Activity
- Referencia:470111-838
- Nº de proveedor:10-500
The Analysis of Minerals and Soils Lab Activity teaches students to utilize physical and chemical techniques to thoroughly investigate crime scene samples.
- For Grades Nine through 12
- Analyses Include Comparison of Acidity and Comparison of Particle Sizes Using Sieves
- Students Utilize Physical and Chemical Techniques to Investigate Crime Scene Samples
Soil, glass, dust, and specks of metal are just some of the trace evidence analyzed in this kit to link a suspect to the scene of the crime.
Analyses include comparison of acidity, comparison of particle sizes using sieves, comparison of particle densities using density gradient tubes, and comparison of minerals present through the use of wet chemistry. A fume hood; a UV light; a balance; a microscope, oven, lamp, or hot plate; glassware; and general lab supplies are required but not included.