Eisco® Amoeba Proteus Model
Amoeba Proteus, 1, 25 cm, 34 cm, 19 cm
Catalog # 470111-026
Supplier: VWR International
Jump to:
Product Details & Documents
This model reveals all the critical parts of the Amoeba proteus.
- Locomotive Position
- Model Mounted on Base
- Movement of Food Vacuole Demonstrated
It's in a locomotive position and mounted on a base.
The model shows the nucleus, contractile vacuole, plastids, and more. It also reveals the movement of food vacuole.
Specifications
- Description:Amoeba Proteus
- Height:19 cm
- Length:34 cm
- Width:25 cm
- No. of Parts:1