Chlorine/Ammonia Neutralizer
Ammonia/chlorine deoxifier, 8 oz.
Catalog # 470021-340
Supplier: PET WORLD INC SE
Número de proveedor #: AMMOLOC45D
Jump to:
Product Details & Documents
Simplify aquarium setup.
- Treats up to 480 gallons
- Use for setup or water changes
Neutralize ammonia produced by fish waste, and remove chlorine and chloramine from tap water, with one easy-to-use treatment. It works in freshwater or saltwater aquariums.
Especificaciones
- Description:Ammonia/chlorine deoxifier
- Size:8 oz.