Alloy Tuning Fork Sets
Supplier: VWR International
Jump to:
Acerca de este artículo
Two sets of popular tuning forks are available.
- Useful Selection of Frequencies
- Two Sets Available
- Setting from A to G
One set features four tuning forks (256 C, 320 E, 384 G, 512C).
The other set features eight tuning forks (256 C, 320 E, 384 G, 480 B, 288 D, 341.3 F, 426.7 A, 512 C).
2 Options Available Below
Product Details & Documents
Two sets of popular tuning forks are available.
- Useful Selection of Frequencies
- Two Sets Available
- Setting from A to G
One set features four tuning forks (256 C, 320 E, 384 G, 512C).
The other set features eight tuning forks (256 C, 320 E, 384 G, 480 B, 288 D, 341.3 F, 426.7 A, 512 C).
Product Family Options
Información del producto
- AvailabilityPrice
- 470148-640Tuning Forks, Set of 4Tuning Forks, Set of 4Kit de 1$285.91Case de 25 (1UN)$311.64
- 470104-594Tuning Forks, Set of 8