Pulley
Single sheave
Catalog # 470049-642
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: PULS01
Jump to:
Product Details & Documents
Smooth running pulley for investigating mechanical advantage.
- V grooves
- Swivel hooks
These free-running, sturdy pulleys feature 50mm diameter plastic sheaves with deep V-grooves, aluminum frames, and swivel hooks on both ends.
Especificaciones
- Description:Single sheave