Glass Microfiber Filter Paper
Glass Microfiber Filter Paper
Catalog # 470201-466
Supplier: AHLSTROM MUNKSJO FILTRATION LLC.
Supplier #: 1610-1100
Jump to:
Product Details & Documents
The glass filter paper is excellent for microwave testing where temperatures get very high such as for use in a food experiment where all the moisture is microwaved out to see how much solid is left behind. Withstands temperatures to 500 °C. Size of each: 11 cm diameter.
Packaging: Package of 100.
Specifications
- Description:Glass Microfiber Filter Paper