A-Frame Base
A-Frame Base, Small
Catalog # 470121-238
Supplier: VWR International
Jump to:
Product Details & Documents
This cast iron base features threads for rods sized 1/2 - 13.
Legs are 21 cm long on the small base and 25 cm long on the large base.
Especificaciones
- Description:A-Frame Base, Small
Accessories
Información del producto
- Catalog #AvailabilityPrice
- Aluminum Rod, 24"L, 1/2"dia, Thread Size: 1/2-13Aluminum Rod, 24"L, 1/2"dia, Thread Size: 1/2-13470016-294Log in Para disponibilidadEach (1UN) - $149.24