Orders LinkContactUsLinkComponent
Adipose Tissue Slide
Adipose Tissue, Section, Osmium
Catalog # 470182-628
Supplier:   Avantor
Adipose Tissue Slide
Catalog # 470182-628
Supplier:  Avantor
Adipose Tissue Slide
Adipose Tissue Slide
Adipose Tissue, Section, Osmium
Price
$193.60
Log in for availability
Each (1UN)
Product Details & Documents
Stores Energy and Insulates the Body
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Fat is retained in cells and treated with osmium tetroxide.
Specifications
  • Description:
    Adipose Tissue
  • Stain:
    Osmium
  • Orientation:
    Section