Acute Lymphatic Leukemia Slide
Acute Lymphatic Leukemia, Smear, Giemsa
Catalog # 470183-236
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates abnormal white blood cells
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Shows abnormal lymphocytes with cleaved or blastic nuclei.
Especificaciones
- Description:Acute Lymphatic Leukemia
- Stain:Giemsa
- Orientation:Smear