Acrylic Ray Optics Set
Acrylic Ray Optics Set
Catalog # 470149-474
Supplier: VWR International
Jump to:
Product Details & Documents
A Good Optics Set for Demonstrations or First Experiments
- Made of polished acrylic
- Provided in a convenient storage box
Set consists of: rectangle (75mm slides), double convex lens and two double concave lenses with a focal length of about 10cm, equilateral prism, and semi-circular block. 15mm thick.
Especificaciones
- Description:Acrylic Ray Optics Set