Acrylic Bell Jar
Acrylic Bell Jar
Catalog # 470121-432
Supplier: VWR International
Jump to:
Product Details & Documents
Lasts for Years!
- Bell operates on 6–12 volts.
Made of tough resistant acrylic, this bell jar is durable enough for years of student use. It is supplied with a rubber stopper equipped with 4mm terminals for input to the bell.
Especificaciones
- Description:Acrylic Bell Jar