Orders LinkContactUsLinkComponent
Acetate Sheet, 8 1/2×11" Each
Acetate Sheet, 8 1/2×11" Each
Acetate Sheet, 8 1/2×11" Each
Certificates
Acetate Sheet, 8 1/2×11" Each
Acetate Sheet, 8 1/2×11" Each
1 Options Available Below
Product Details & Documents
Acetate Sheet, 8 1/2 x 11" Each
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Product Family Options
Información del producto
  • Availability
    Price
  • 470003-608
    Plastic Strip, Clear Acetate, 1" x 10"0
    Plastic Strip, Clear Acetate, 1" x 10"0
    Discontinued
    Supplier Number: -
  • 470161-656
    Acetate Sheet, 8 1/2 x 11" Each
    Acetate Sheet, 8 1/2 x 11" Each
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4.82
    $4.82
    Log in Para disponibilidad
    Caja de 100 (1UN)
    Caja de 100 (1UN) - $5.25
    $5.25
    Each (1UN)
       $4.82
    Caja de 100 (1UN)
       $5.25
    Supplier Number: 5199193