Conductivity Indicator
Supplier: SCIENCE INTERACTIVE GROUP
Jump to:
About this item
Light Bulb Glows When Water Conducts Electricity.
- Attaches to a standard ring stand
- Comes with illustrated instructions
Presenting no shock hazard because the cord at the side can’t come in contact with the water, this indicator features a plastic molded body and recessed electrical connections and electrodes. It operates on 110 VAC. A light bulb is required.
2 Options Available Below
Product Details & Documents
Light Bulb Glows When Water Conducts Electricity.
- Attaches to a standard ring stand
- Comes with illustrated instructions
Presenting no shock hazard because the cord at the side can’t come in contact with the water, this indicator features a plastic molded body and recessed electrical connections and electrodes. It operates on 110 VAC. A light bulb is required.
Product Family Options
Product Information
- AvailabilityPrice
- 470016-796Conductivity IndicatorSupplier Number: 611-2260
- 470061-754Bulb (15W) for Water ConductivitySupplier Number: 674-0035