Bulb socket board
Supplier: VWR International
Jump to:
About this item
This solid socket board holds light bulbs for circuit experiments.
- Choice of Three or Five-Socket Sizes
- Removable Shunt Wires Featured on Five-Socket Size
- Terminals Featured on Three-Socket Board
- Plastic Bases with Rubber Feet
- Small Incandescent Bulbs
It's available in three or five-socket sizes. Terminals on three-socket board let users connect lamps in series or parallel, and five-socket board features removable shunt wires for the same purpose.
All units have plastic bases with rubber feet, and small incandescent bulbs are included.
2 Options Available Below
Product Details & Documents
This solid socket board holds light bulbs for circuit experiments.
- Choice of Three or Five-Socket Sizes
- Removable Shunt Wires Featured on Five-Socket Size
- Terminals Featured on Three-Socket Board
- Plastic Bases with Rubber Feet
- Small Incandescent Bulbs
It's available in three or five-socket sizes. Terminals on three-socket board let users connect lamps in series or parallel, and five-socket board features removable shunt wires for the same purpose.
All units have plastic bases with rubber feet, and small incandescent bulbs are included.
Product Family Options
Product Information
- AvailabilityPrice
- 470149-514Three-SocketThree-SocketEach (1UN)$337.51Case de 25 (1UN)$367.88
- 470149-512Five-SocketFive-SocketEach (1UN)$408.47Case de 15 (1UN)$445.23