Orders LinkContactUsLinkComponent
Ward's® 6" Photo Rules
Ward's® 6" Photo Rules
Supplier:   OREGON RULE CO.
Ward's® 6" Photo Rules
Supplier:  OREGON RULE CO.
Certificates
Ward's® 6" Photo Rules
Ward's® 6" Photo Rules
2 Options Available Below
Product Details & Documents
Select the Right Contrast Ruler
  • White or Black Available
These plastic rules are graduated in 1/16" and 1 mm increments. Select black or white for the appropriate
contrast required for photography.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 470001-834
    Photo Rule, White
    Photo Rule, White
    6"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $29.32
    $29.32
    Each (1UN)
       $29.32
    Supplier Number: 4RFM-WH006PL/WARDS
  • 470001-836
    Photo Rule, Black
    Photo Rule, Black
    6"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $29.32
    $29.32
    Each (1UN)
       $29.32
    Supplier Number: 4RFM-BK006PL/WARDS