Hexagram Weight Set
20 x 1 g, 20 x 5 g, 10 x 10 g, 4 x 20 g, 2 x 100 g, 1 x 500 g, 1 x 1000 g Masses
Catalog # 470046-280
Supplier: LEARNING RESOURCES SE
Número de proveedor #: 32071
Jump to:
Product Details & Documents
58 Colorful Metric Weights.
- Stackable color coded weights
- Plastic storage box included
These color coded metric weights are perfect for your classroom. A total of 58 pieces totals 2000g.
Caution: CHOKING HAZARD- Small Parts. Not for Children Under 3 Years.
Especificaciones
- Includes:20 x 1 g, 20 x 5 g, 10 x 10 g, 4 x 20 g, 2 x 100 g, 1 x 500 g, 1 x 1000 g Masses