Three-Meter Measuring Tape
Measuring tape, metric and imperial, 3 m (10 ft.)
Catalog # 470020-044
Supplier: AMERICAN SCIENTIFIC LLC SE
Número de proveedor #: 8777-30
Jump to:
Product Details & Documents
Perfect for a variety of small applications, the bright yellow tape retracts neatly into a compact, high-impact plastic case.
- Aligned with GLOBE protocols
Graduated in ¹/₁₆" and 1 mm increments.
Especificaciones
- Description:Measuring tape, metric and imperial
- Length:3 m (10 ft.)