3B Scientific® Pelvic Skeleton Models
Pelvis - Male, 1, 25 cm, 16 cm, 21 cm
Catalog # 470030-162
Supplier: AMERICAN 3B SCIENTIFIC SE
Número de proveedor #: 1000133
Jump to:
Product Details & Documents
The 3B Scientific® pelvis skeleton replicas consists of hip bone, sacrum with coccyx and 2 lumbar vertebrae. These high-quality anatomical models area great addition to any anatomy classroom or doctor's office.
- Includes 3B smart anatomy
This skeleton is great for detailed anatomical study of the male or female pelvis.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.
Documents
TDS 3B Smart Anatomy
Hoja de datos
Especificaciones
- Description:Pelvis - Male
- Height:21 cm
- Length:16 cm
- Width:25 cm
- No. of Parts:1