Boreal2 Digital Compound Microscopes - HM Series
HM series Standard, 640×480 camera, monocular, LED cordless
470014-544
: DM-SK102
These microscopes have all the advantages of the Boreal2 microscopes plus digital live image display, capture and manipulation.
- Compatible with Windows 7 and above and Mac OSX and above
- Motic image editing software included
- One-year warranty on camera
- Lifetime warranty on microscope chassis
These digital microscopes are both PC and Mac compatible, and the USB connection makes set up easy. Users can capture video or still images and even measure images on their screen. An auto-capture feature lets users collect images at pre-set intervals over time, which is perfect for prolonged class experiments.
Users can also export their results or save them for editing and annotating later. The exported images are compatible with other programs. Three different microscope models with different levels of resolution are available.
: This product includes a manufacturer's warranty.
- Magnification:40X, 100X, 400X
- Description:HM series Standard, 640×480 camera, monocular
- Illumination:LED cordless