Orders LinkContactUsLinkComponent
Boreal2 Microscopes, HM Series
Boreal2 Microscopes, HM Series
Boreal2 Microscopes, HM Series
Certificates
Boreal2 Microscopes, HM Series
3 Options Available Below
Product Details & Documents
With all-metal construction, a rugged, student-proof design, and precision optics, Boreal2 HM Microscopes are ideal for middle and high school students.
  • Rotatable, 45°-Inclined Viewing Head
  • Parfocal and Parcentered Optics
  • Lifetime Manufacturer's Warranty
  • Designed for Middle and High School Students
  • Total Magnification of 400x
A bit more advanced than the EM series, this microscope is designed for middle and high school students. Features include a gliding stage with one-touch stage clips, recessed eyepiece screws to prevent the disassembly of the microscope, a removable cord holder for easy storage, tension control to halt stage drift, large and easy-to-read magnification labels, and a reversed nose piece to protect objectives.
This microscope also features both a coarse and fine focus, has bright, white LED lighting, and offers a total magnification of 400x.
Ordering information: This product includes a manufacturer's warranty.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 470016-038
  • 470014-512
  • 470014-514
  • Objectives:
  • 4×, 10×, 40×
  • 4×, 10×, 40×
  • 4×, 10×, 40×
  • Stage:
  • Gliding
  • Gliding
  • Gliding
Especificaciones
Product Family Options
Información del producto
  • Illumination
    Availability
    Price
  • 470016-038
    Monocular
    Monocular
    LED corded
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $6,929.11
    $6,929.11
    Each (1UN)
       $6,929.11
    Supplier Number: SK101
    Especificaciones:
    More
    Specifications
    Cat. No.
    470016-038
    Objectives
    4×, 10×, 40×
    Stage
    Gliding
  • 470014-512
    Monocular
    Monocular
    LED cordless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $6,609.30
    $6,609.30
    Each (1UN)
       $6,609.30
    Supplier Number: SK102
    Especificaciones:
    More
    Specifications
    Cat. No.
    470014-512
    Objectives
    4×, 10×, 40×
    Stage
    Gliding
  • 470014-514
    Dual View
    Dual View
    LED corded
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $8,101.73
    $8,101.73
    Each (1UN)
       $8,101.73
    Supplier Number: SK110
    Especificaciones:
    More
    Specifications
    Cat. No.
    470014-514
    Objectives
    4×, 10×, 40×
    Stage
    Gliding
Accessories
Información del producto
  • Catalog #
    Electrical
    Availability
    Price
  • Replacement bulb
    Replacement bulb
    470005-050
    1 W
    Log in Para disponibilidad
    Each (1UN) - $159.92