Orders LinkContactUsLinkComponent
Boreal2 Microscopes, HM Advanced Series
Boreal2 Microscopes, HM Advanced Series
Boreal2 Microscopes, HM Advanced Series
Certificates
Boreal2 Microscopes, HM Advanced Series
4 Options Available Below
Product Details & Documents
Perform quantitative experiments over a wide frequency range with these microscopes.
  • Objectives: 4x, 10x, 40x, and 100x
  • Large Magnification Labels
  • Protection against Disassembly
  • Tension Control
  • Charger Included
Bright, white, cold LED lighting and coated-glass optics give excellent images. The 4x, 10x, 40x, and 100x objectives have a wide color band with large, easy-to-read magnification labels on two sides for simplified viewing. Full-size, stable bodies hold a host of student-proof features, including locked-on stage clips, eye pieces, and objectives that offer protection against disassembly.
A four-hole reversed nosepiece protects objectives, and tension control stops stage drift. The microscopes are available in corded or cordless models; a removable cord holder is included on corded models for easy storage. The cordless model can be used anywhere for up to 50 hours with an eight-hour charge; a charger is included.
Ordering information: This series includes a lifetime warranty.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 470018-032
  • 470014-520
  • 470014-518
  • 470014-528
  • Objectives:
  • 4×, 10×, 40×, 100×
  • 4×, 10×, 40×, 100×
  • 4×, 10×, 40×, 100×
  • 4×, 10×, 40×, 100×
  • Stage:
  • Built-In Mechanical
  • Built-In Mechanical
  • Built-In Mechanical
  • Built-In Mechanical
Especificaciones
Product Family Options
Información del producto
  • Illumination
    Availability
    Price
  • 470018-032
    Binocular
    Binocular
    LED corded
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $16,523.22
    $16,523.22
    Each (1UN)
       $16,523.22
    Supplier Number: SK123BINOA
    Especificaciones:
    More
    Specifications
    Cat. No.
    470018-032
    Objectives
    4×, 10×, 40×, 100×
    Stage
    Built-In Mechanical
  • 470014-520
    Monocular
    Monocular
    LED cordless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $10,500.25
    $10,500.25
    Each (1UN)
       $10,500.25
    Supplier Number: SK122
    Especificaciones:
    More
    Specifications
    Cat. No.
    470014-520
    Objectives
    4×, 10×, 40×, 100×
    Stage
    Built-In Mechanical
  • 470014-518
    Monocular
    Monocular
    LED corded
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $8,794.62
    $8,794.62
    Each (1UN)
       $8,794.62
    Supplier Number: SK121
    Especificaciones:
    More
    Specifications
    Cat. No.
    470014-518
    Objectives
    4×, 10×, 40×, 100×
    Stage
    Built-In Mechanical
  • 470014-528
    Dual View
    Dual View
    LED cordless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13,538.37
    $13,538.37
    Each (1UN)
       $13,538.37
    Supplier Number: SK132
    Especificaciones:
    More
    Specifications
    Cat. No.
    470014-528
    Objectives
    4×, 10×, 40×, 100×
    Stage
    Built-In Mechanical
Accessories
Información del producto
  • Catalog #
    Electrical
    Availability
    Price
  • Replacement bulb
    Replacement bulb
    470005-050
    1 W
    Log in Para disponibilidad
    Each (1UN) - $159.92
Información del producto
  • Catalog #
    No. of Slides
    Type
    Availability
    Price
  • Advanced Histology Slide Set
    Advanced Histology Slide Set
    470177-704
    50
    Human Body
    Log in Para disponibilidad
    KIT(1UN) - $11,663.83