Orders LinkContactUsLinkComponent
Twistit® Rubber Stoppers
Twistit® Rubber Stoppers
Twistit® Rubber Stoppers
Certificates
Twistit® Rubber Stoppers
2 Options Available Below
Product Details & Documents
An outstanding advance in rubber stoppers, four stoppers in one reduce inventory confusion and cost.
  • Twist one of the rubber protrusions off and the solid stopper becomes a one-hole stopper
  • Twist the second one off for a two-hole stopper
  • Twist the third one off for a three-hole stopper
Made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation.
Product Family Options
Información del producto
  • Availability
    Price
  • 470003-668
    Stopper, Rubber, Twistit, Size 7, Black, 3 Holes
    Stopper, Rubber, Twistit, Size 7, Black, 3 Holes
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 40
    Paquete de 40 - $1,155.51
    $1,155.51
    Log in Para disponibilidad
    Case de 10 (40UN)
    Case de 10 (40UN) - $1,259.50
    $1,259.50
    Paquete de 40
       $1,155.51
    Case de 10 (40UN)
       $1,259.50
    Supplier Number: J7--M299
  • 470200-224
    Stopper, Rubber, Twistit, Size 4, Black, 2 Holes
    Stopper, Rubber, Twistit, Size 4, Black, 2 Holes
    Discontinued
    Supplier Number: G4--M299
  • 470201-232
    Stopper, Rubber, Twistit, Size 6, Black, 3 Holes
    Stopper, Rubber, Twistit, Size 6, Black, 3 Holes
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,145.96
    $1,145.96
    Each (1UN)
       $1,145.96
    Supplier Number: I6--M299