Dissectible Student Leyden Jar
Catalog # 470002-944
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Descripción:Jar, Leyden
- Referencia:470002-944
- Nº de proveedor:LYJ001
Specifications
About this item
Quickly demonstrate the isolation of a static charge.
- Easily assembles or disassembles
- Large electrode for attracting a charge
Learn about static electricity and the function of a dielectric with this unique device. Two aluminum jars - one with an attached aluminum electrode and one plastic jar – are used to safely collect the charge.