NMC Write-On NFPA Labels
Write-on NFPA labels, PS Paper, 50.8×50.8 mm (2×2")
Catalog # 56606-750
Supplier: ACCUFORM MANUFACTURING, INC
Supplier #: HMS12L
Jump to:
Product Details & Documents
Write-on labels allow you to customize your message.
Specifications
- Description:Write-on NFPA labels
- Dimensions:50.8×50.8 mm (2×2")
- Material:PS Paper