Black Dragon Zero Powder-Free Nitrile Exam Gloves, Microflex®
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificates
About this item
Black nitrile gloves with textured fingertips offer better tactile sensitivity and grip.
- Ambidextrous
- Powder-free
- Beaded cuffs
Unique black color masks stains and acts as a backdrop for powders, solvents, etc.
Packaging: Packaged 1,000 gloves per case, 100 gloves per dispenser box by weight.
Specifications
- Catalog No:
- 89230-608
- 89230-610
- 89230-600
- 89230-602
- 89230-604
- 89230-606
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Desechable
- Espesor del puño:
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- Espesor del dedo:
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- Espesor de la palma:
- 3.5 mil
- 3.5 mil
- 3.5 mil
- 3.5 mil
- 3.5 mil
- 3.5 mil