Vial Connecting Adapters, PTFE, Ace Glass Incorporated
Catalog # 89077-628
Supplier: ACE GLASS, INC.
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Referencia:89077-628
- Fits:Adapters 89077-630, 89082-468
- Grosor:22-400
- Ace-thred no.:#15
- Nº de proveedor:13290-22
Specifications
About this item
Chemically inert PTFE adapters feature either a #15 or #25 Ace-Thred.
- GPI vial inner thread
- Suitable for vacuum work
- O-ring included
A GPI vial inner thread connects conversion adapters to the matching vial for use in rotary evaporators. Suitable for vacuum work. Supplied with either FETFE or Chemraz O-ring. Thread Size represents the inner GPI Thread.
Caution: FETFE is not suitable for use with methylene chloride or acetone. Chemraz is suitable for use instead.