Scribes, Diamond Tipped, Excelta Corp®
Catálogo # 94024-264
Proveedor: EXCELTA CORPORATION
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Especificaciones
- Point Size:0.5×0.5 mm (0.02×0.02" )
- Descripción:Aluminum scribe, diamond tipped, 3 star quality
- Longitud:112.5 mm (4.5" )
- Referencia:94024-264
- Nº de proveedor:475A
Especificaciones
Acerca de este artículo
Diamond coated tips are very strong.
- Light weight aluminum handle
- Ideal for scraping
These diamond tipped scribes with aluminum handles and 20 mil fine tip are ideal for scraping.