Tweezers, Broad Flat Wafer Handling, EXCELTA®
Catalog # 63042-026
Supplier: EXCELTA CORPORATION
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Descripción:Tweezers, flat point, 1 star quality
- Matriz:Stainless steel, anti-magnetic
- Curvature:Straight
- Referencia:63042-026
- Estilo de la punta:Straight Flat Tip
- Longitud total:118.75 mm (4.75")
- Nº de proveedor:35A-SA-SE
Specifications
About this item
These were the original wafer tweezers. They feature straight broad flat tips.
- Large flat tip offers better control of item
- Anti-magnetic stainless steel, cleanroom safe
- Two different quality grades
These tweezers feature straight large flat round pointed tips and serrated handles for better control. They are made of anti-magnetic Neverust® stainless steel and is 4.75" long. We offer 2 different qualities. The higher the quality, the more precise the tips.