Four Star™ Tweezer, Straight Copolymer Soft Replaceable Tip, EXCELTA®
94024-140
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Descripción:Tweezers, replaceable tip, 4 star quality
- Longitud:12.7 cm (5"), Overall
- Matriz:Copolymer tips
- Tip Dimensions:.020"
- Curvature:Straight
- Referencia:94024-140
- Estilo de la punta:Straight Tip
- Longitud total:128 mm (5.125")
- Nº de proveedor:179A-RT
- Tip Width:0.02" (0.5 mm)
- Tip Length:0.25 mm (0.010")
These replaceable tip tweezers utilize a copolymer material that is ESD Safe (10 to the 10th Ohm) and temperature resistant to 185°F.
- Static dissipative for work with sensitive electronic work
- Cleanroom safe
- Soft tips won't scratch or mar
The Copolymer replaceable tip tweezers are static dissipative with a surface resistivity is (10⁶ to 10⁷ ohms/sq). The soft copolymer tips are ESD safe (10⁶ to 10⁷ ohms/sq), temperature resistant to 195 °F (90 °C), and silicone/chloride free. They are the ideal tweezers for work in the cleanroom. The body is made of anti-magnetic Neverust® stainless steel (AISI 316L). All replacement tips come complete with 2 tips and 2 screws. They are excellent for applications requiring no metal-to-metal contact, avoiding scratching and marring.