ContactUsLinkComponent
PHCbi MDF Series –30 °C Biomedical Upright Freezers, PHC Corporation
PHCbi MDF Series –30 °C Biomedical Upright Freezers, PHC Corporation
Catalog # 89429-298
Proveedor:  PHC Corporation
Product available on GSA Advantage®
undefined
PHCbi MDF Series –30 °C Biomedical Upright Freezers, PHC Corporation
Catalog # 89429-298
Proveedor:  PHC Corporation
Supplier Number:  MDF-U731-PA
Product available on GSA Advantage®
Especificaciones
  • Rango de temperatura:
    –15...–30 °C (+5...–22 °F)
  • Capacidad:
    623 L (22 cu.ft.)
  • Descripción:
    MDF series –30 °C Biomedical upright freezer with automatic defrost fits 16× side access racks or sliding drawer racks
  • Modelo:
    MDFU731PA
  • Referencia:
    89429-298
  • Detalles eléctricos:
    115 V, 60 Hz, 15 A, NEMA 5 - 15P plug
  • Estantes/cajones:
    4 adjustable wire shelves, 50 kg (110 lbs.) capacity each
  • Peso:
    152 kg (335 lbs.)
  • Aislamiento:
    CFC-free polyurethane foamed-in place
  • Exterior W×D×H:
    770×830×1955 mm (30.3×32.7×77")
  • Interior W×D×H:
    650×700×1370 mm (25.6×27.5×53.9")
  • Nº de proveedor:
    MDF-U731
Especificaciones
Acerca de este artículo
PHCbi MDF Series biomedical freezers are designed for short- or intermediate-term storage at temperatures as low as –30 °C.
  • For intermediate storage
  • Uniform temperature maintenance
  • Built-in microprocessor
  • Two-year manufacturer's parts and labor warranty
  • Three-year manufacturer's compressor parts warranty
Constructed with high-performance laboratory and clinical-grade refrigeration systems, these freezers are used in medical, biotechnology, and industrial laboratories for storage of blood components, enzymes, culture media, reagents and specimens.
Uniform temperatures are maintained throughout the chamber using shelf freezing construction. The PHCbi-built microprocessor controller, with digital display of all input/output functions, provides an intuitive method to maintain set point, alarm monitoring, and diagnostic functions. PHCbi’s freezing coils within the shelf offer exceptional temperature recovery due to the expanded surface area offered by the shelf freezing design. Rigid polyurethane foam insulation.
Ordering information: All units are supplied with a two-year manufacturer's parts and labor warranty and a three-year manufacturer's compressor parts warranty. Bins and shelves are sold separately.
Temperature Chart Recorders
Información del producto