About this item
The PHCbi MIR series offers accurate temperature control and uniformity over a wide temperature range, making it suitable for various applications.
- Accurate temperature control
- Wide temperature range and uniformity
- Microprocessor, 8 bit
- Two-year manufacturer's parts and labor warranty
- LCD controller
These incubators are exceptional units for a wide range of applications requiring a temperature range of –10 to 60 °C
The microprocessor, combined with a heater P.I.D. and compressor On-Off system, allows for precision temperature control. With a feed forward function that inputs the operating conditions of the compressor beforehand, the system ensures accurate temperature control for the chamber. The new LCD controller features a pop-up menu that displays current conditions (temperature and date), alarm conditions, door and light status, and various settings for each program (12 step, 10 program, and more).
Programmable models include three-step functions useful for investigations involving microbiology, plant cell biology, and more. The MIR Series incorporates an 8-bit microprocessor controller for heat and refrigeration control (±0.2 °C). The simple program input accommodates a range of diversified experimentation requirements. It is ideal for applications that require settings to be changed, microorganism cultures and preservation, and experimentation during night time or holidays.
Specifications
- Catalog No:
- 97015-590
- 97015-588
- 97049-050
- Aislamiento:
- Rigid Polyurethane, Foamed-In Place
- Rigid Polyurethane, Foamed-In Place
- Rigid Polyurethane, Foamed-In Place
- Detalles eléctricos:
- 115V, 60Hz, 1 Phase
- 115V, 60Hz, 1 Phase
- 115V, 60Hz, 1 Phase