Nalgene® Lab Carrier, Thermo Scientific
Catálogo # 16190-903
Proveedor: Thermo Fisher Scientific
Especificaciones
- Descripción:HDPE lab carrier
- Referencia:16190-903
- La x An x Al:38×36×27 cm (15×14×50½")
- Nº de proveedor:7131-0001
Especificaciones
Acerca de este artículo
Nalgene Lab Carriers reduce breakage and spillage when transporting glass and plastic bottles
They are made of sturdy, High Density polyethylene construction and will hold six full 1000mL glass media bottles. The carriers also stack when they are empty.