Molecular BioProducts Low Retention Pipette Tips, Thermo Scientific
Catálogo # 72830-428
Proveedor: MOLECULAR BIO-PRODUCTS INC
Especificaciones
- Volumen:0.1 - 10 µl
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:ISO14001 (Certificate Registration No. EMS 752603)
- Referencia:72830-428
- Sterile:+
- Estilo de la punta:Pure 10, Reach
- Nº de proveedor:3511-05
Especificaciones
Acerca de este artículo
Molecular BioProducts low retention pipette tips are specifically designed for eliminating sample retention within the pipette tip.
All Pure tips are racked, pre-sterilized, and certified free of RNase, DNase, DNA, and pyrogen.
The inner surface of the tip is extremely hydrophobic, reducing loss of sample from adhesion to the tip interior. Tips are crystal clear to allow easy sample viewing.