Especificaciones
- Volumen:1000 µl (1 ml)
- Max. temperature:50 °C / 122 °F
- Descripción:Model 1001 LTN CTC Syringe
- Min. temperature:20 °C / 68 °F
- Needle length:2 inch (51 mm)
- Esterilizable en autoclave:No
- Esterilizable por gas:Yes
- Point style:3
- Referencia:89173-256
- Calibrador:22 gauge
- Nº de proveedor:203081
- Standard Needle Dead Volume:6.81 μl
- Termination:LTN
Especificaciones
Acerca de este artículo
Hamilton offers 21 quality syringe options, S-Line, C-Line and X-Type, for the CTC/Leap LC/HTC/HTS Pal, and 2 quality syringe options for the CTC/Leap A200S HPLC Autosampler.
Hamilton maintains a large catalog of replacement syringes for the most popular HPLC autosamplers. Our syringes are handmade with an unmatched attention to detail. The manufacturing process and quality assurance procedures ensure that every syringe we sell will provide superior accuracy and precision as well as cycle life. Choose the sub category that is associated with your autosampler manufacturer to find the syringes available for your instrument.
Syringes in this category are either 700 series Microliter™ or 1700 and 1000 series Gastight® syringes. Microliter™ syringes have a stainless steel plunger that is hand fitted to the glass barrel and are designed for liquid sampling. Gastight® syringes have a PTFE plunger tip that creates tight connection between the syringe barrel and plunger allowing for liquid or gas sampling.
Cemented Needle (N)- For low volume syringes, the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and is not user-selectable. Removable Needle (RN)- The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life. Luer Tip Cemented Needle (LTN)- For mid volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable. Fixed Needle (FN) - This termination is found on CTC chromatography syringes. The unique design features a direct attachment of the needle to the barrel and eliminates contact between the sample solvent and the adhesive reducing carryover. Needle Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting.