ContactUsLinkComponent
Condenser, Allihn, with No. 7 Ace-Thred Connections
  89053-920
Product available on GSA Advantage®
Small Business Enterprise
undefined
Condenser, Allihn, with No. 7 Ace-Thred Connections
  89053-920
 :  5946-118
Product available on GSA Advantage®
Small Business Enterprise
  • Descripción:
    Condenser, Allihn, "Ace-Safe" Connectors
  • Referencia:
    89053-920
  • Top ST Joint Size:
    [ST] 24/40
  • Jacket Length:
    300 mm
  • Nº de proveedor:
    5946-118
Bulb type Allihn condenser, approximately one bulb per 50 mm, with inner and outer joint at bottom and top
Fabricated from borosilicate glass. No.7 Ace-Thred hose connections on inlet and outlet with barb for 1/4-inch I.D. tubing.
 : Available with or without "Ace-Safe" Connectors.