FreeZone® Product Shelves, Labconco®
Product Shelves fit inside FreeZone® Clear or Port Drying Chambers to accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications.
- Removable, 10" Ø aluminum shelves with ¼" rim slide into the stainless steel support
- Product shelves are designed to fit inside drying chambers
- Shelf can be placed in freezer to pre-freeze sample for quick loading
- The removable shelf design allows for flexible shelf height spacing
Also available are heated shelves that include ambient to +40 °C (+104 °F) variable heat control on models manufactured 2016 and earlier and up to +60 °C (+140 °F) on models 2017 and later (contact your Avantor sales representative for more information).
: Includes either three or five removable, 10" Ø aluminum shelves with ¼" rims, one stainless steel support and three rubber feet. Drying chambers and freeze dryers sold separately.
- Catalog No:
- 75840-998
- Environmentally Preferable:
- Sustainable Materials
Frequently Bought Together