Blower Transition Adapters, Labconco®
Blower Transition Adapter, 30.5 cm (12"), 1.8 kg (4 lbs.)
Catalog # 30186-002
Supplier: LABCONCO CORPORATION MX
Número de proveedor #: 7003400
Jump to:
Product Details & Documents
Epoxy-coated steel blower transition adapters fit all Labconco coated steel blowers
Adapter allows you to connect thermoplastic duct to the exhaust side of a blower to create an exhaust stack. A nominal size PVC duct fits inside the opening.
Especificaciones
- Description:Blower Transition Adapter
- Shipping Weight:1.8 kg (4 lbs.)
- Nominal Diameter:30.5 cm (12")