Black Phenolic Dropper Assembly, Qorpak®
7×66 mm, 24-400
Catalog # 470147-506
Supplier: QORPAK
Supplier #: 502253
Jump to:
Product Details & Documents
Dropper assemblies feature a glass tube with black polypropylene closure and rubber squeeze bulb
Tip opening has a 3.0 mm outside diameter.
Documents
Qorpak Laboratory Container and Supply
Product Literature
Especificaciones
- Cap Size:24-400
- Size:7×66 mm