Orders LinkContactUsLinkComponent
Nalgene® Multi-Bottle Rack; White PC, Thermo Scientific
Nalgene® Multi-Bottle Rack; White PC, Thermo Scientific
8 compartment carrier for 500 ml bottles
Catalog # 16161-850
Supplier #:  DS5996-0871
Nalgene® Multi-Bottle Rack; White PC, Thermo Scientific
Catalog # 16161-850
Nalgene® Multi-Bottle Rack; White PC, Thermo Scientific
Nalgene® Multi-Bottle Rack; White PC, Thermo Scientific
8 compartment carrier for 500 ml bottles
Price
$3,440.00
Log in for availability
Each (1UN)
Product Details & Documents
Nalgene White Polycarbonate Multi-Bottle Rack holds up to eight 500mL round or square bottles securely
The bottles fit snugly between upper grid and molded-in depression on lower tray. The multi racks contain handle telescopes for convenient storage.
Specifications
  • Description:
    8 compartment carrier for 500 ml bottles