Round Winpaks®, High-Density Polyethylene, Qorpak®
Catalog # 11311-368
Supplier: QORPAK
Small Business Enterprise
Specifications
- Tamaño de la tapa:70-400
- Capacidad:19 L (5 gal.)
- Color:Natural
- Descripción:Natural Polyethylene Gasket Lined
- Matriz:HDPE
- Referencia:11311-368
- Nº de proveedor:PLA-03327
Specifications
About this item
Winpaks® are cylindrical in shape with spout placement in line with the sidewall for maximum removal of product during dispensing.
The unit is also equipped with a swing handle on the top face. Designed for the secure, dependable shipment and storage of liquid products.