Orders LinkContactUsLinkComponent
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Supplier:   QORPAK
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Supplier:  QORPAK
Certificates
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Sample Vials, Clear Borosilicate Glass, Screw-Thread
7 Options Available Below
Product Details & Documents
Clear, flint screw-thread sample vials offer maximum visibility.
  • Available with a variety of caps, or as vials only, to meet specific requirements
  • Pre-cleaned options save time and money
These general-purpose glass vials are made of Type I Borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.
Product Family Options
Clear borosilicate screw thread vials with green thermoset F217 and PTFE lined cap
Información del producto
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 66009-558
    Vials without treatment
    Vials without treatment
    7.50 ml
    17×60 mm
    Clear
    Yes
    15-425
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (144UN)
    Case (144UN) - $4,227.25
    $4,227.25
    Case (144UN)
       $4,227.25
    Supplier Number: GLC-00984
  • 66009-556
    Vials without treatment
    Vials without treatment
    1.90 ml
    12×35 mm
    Clear
    Yes
    8-425
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (144UN)
    Case (144UN) - $4,244.24
    $4,244.24
    Case (144UN)
       $4,244.24
    Supplier Number: GLC-00978
  • 66009-557
    Vials without treatment
    Vials without treatment
    3.80 ml
    15×45 mm
    Clear
    Yes
    13-425
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (144UN)
    Case (144UN) - $3,557.39
    $3,557.39
    Case (144UN)
       $3,557.39
    Supplier Number: GLC-00980
  • 66009-559
    Vials without treatment
    Vials without treatment
    11.00 ml
    19×65 mm
    Clear
    Yes
    15-425
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (144UN)
    Case (144UN) - $4,803.72
    $4,803.72
    Case (144UN)
       $4,803.72
    Supplier Number: GLC-00988
  • 66009-555
    Vials without treatment
    Vials without treatment
    30.00 ml
    25×95 mm
    Clear
    Yes
    22-400
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (144UN)
    Case (144UN) - $6,715.97
    $6,715.97
    Case (144UN)
       $6,715.97
    Supplier Number: GLC-01008
  • 11311-648
    Vials without treatment
    Vials without treatment
    40.00 ml
    27.75×95 mm
    Clear
    Yes
    24-400
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (144UN)
    Case (144UN) - $8,443.65
    $8,443.65
    Case (144UN)
       $8,443.65
    Supplier Number: GLC-01022
Clear borosilicate screw thread vials with black phenolic polycone lined cap
Información del producto
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 16087-068
    Vials without treatment
    Vials without treatment
    22.00 ml
    27.25×70 mm
    Clear
    Yes
    24-400
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (72UN)
    Case (72UN) - $3,461.71
    $3,461.71
    Case (72UN)
       $3,461.71
    Supplier Number: GLC-01007