ContactUsLinkComponent
OIML Class F1 Precision Weight Sets
OIML Class F1 Precision Weight Sets
Catálogo # 12766-056
Proveedor:  TROEMNER LLC
Product available on GSA Advantage®
undefined
OIML Class F1 Precision Weight Sets
Catálogo # 12766-056
Proveedor:  TROEMNER LLC
Número de proveedor:  80780448
Product available on GSA Advantage®
Especificaciones
  • Referencia:
    12766-056
  • Certificación:
    NVLAP
  • Peso:
    1 kg - 1 g
  • Nº de proveedor:
    7416-F1W
  • No. of Weights:
    13
Especificaciones
Acerca de este artículo
Troemner OIML Calibration Weights conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111)
Weights 1 g and above are two-piece construction. Troemner OIML Calibration Weights 1 g and larger are cylindrical in shape with a tapered neck for easy gripping. They are perfectly flat on top to stack easily on center. Weight bottoms are slightly recessed to expose the smallest possible area to wear.
They are manufactured from the finest stainless steels available including Troemner Alloy 8 Stainless Steel that has low magnetic permeability, uniform consistency and high resistance to corrosion.